Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003124.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003124.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003124.hg.1 |
TC15000347.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009272.hg.1 |
ENST00000558917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009272.hg.1 |
ENST00000558917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009272.hg.1 |
ENST00000558917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009272.hg.1 |
ENST00000558917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009272.hg.1 |
ENST00000558917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007104.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009272.hg.1 |
ENST00000558917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009272.hg.1 |
ENST00000558917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009272.hg.1 |
ENST00000558917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009272.hg.1 |
ENST00000558917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009272.hg.1 |
ENST00000558917.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150474.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150474.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150474.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150474.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150474.hg.1 |
ENST00000340827.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150474.hg.1 |
ENST00000557864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150474.hg.1 |
ENST00000559137.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500150474.hg.1 |
NM_152448.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500150474.hg.1 |
TC1500007104.hg.1 |
Clariom D TC |