Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13010024.hg.1 |
ENST00000356501.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13010024.hg.1 |
ENST00000375418.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13010024.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13010024.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13010024.hg.1 |
TC13000894.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300165958.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165958.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165958.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165958.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165958.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165958.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165958.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165958.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300165958.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165958.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300165958.hg.1 |
TC1300009906.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000356501.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000375418.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
ENST00000413169.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_138430.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009906.hg.1 |
NM_199162.2 |
Refseq |