Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009628.hg.1 |
TC09000749.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009628.hg.1 |
ENST00000357304.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009628.hg.1 |
ENST00000372249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009628.hg.1 |
ENST00000405995.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009628.hg.1 |
ENST00000418650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009628.hg.1 |
ENST00000458550.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09009628.hg.1 |
NM_013318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185103.hg.1 |
ENST00000357304.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185103.hg.1 |
ENST00000451855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185103.hg.1 |
NM_013318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185103.hg.1 |
ENST00000357304.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185103.hg.1 |
ENST00000451855.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900185103.hg.1 |
NM_013318.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900185103.hg.1 |
TC0900012184.hg.1 |
Clariom D TC |