Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
TC08001370.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
ENST00000220669.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
ENST00000517588.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
ENST00000519523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
ENST00000521287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
ENST00000521895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
ENST00000522520.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
ENST00000523096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
NM_001170796.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
NM_001170797.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
NM_024699.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
NR_033193.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
NR_033194.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
NR_033195.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08017710.hg.1 |
NR_033196.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000220669.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000517450.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000517588.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000518419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000519464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000519523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000519820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000520076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000521287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000521895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000522520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000523096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000523431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000524339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NM_001170796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NM_001170797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NM_024699.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NR_033193.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NR_033194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NR_033195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NR_033196.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000220669.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000517450.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000517588.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000518419.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000519464.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000519523.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000519820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000520076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000521287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000521895.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000522520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000523096.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000523431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
ENST00000524339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NM_001170796.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NM_001170797.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NM_024699.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NR_033193.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NR_033194.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NR_033195.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
NR_033196.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800187567.hg.1 |
TC0800012435.hg.1 |
Clariom D TC |