Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01015913_m1
See other PBRM1 GE Assays ›
Gene Symbol:
PBRM1
Gene Name:
polybromo 1
Gene Aliases:
BAF180, PB1
Chromosome Location:
Chr.3: 52545352 - 52685933 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01015913_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: PBRM1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_005265279.4
XP_005265336.1
29 - 30
4840
81
XM_005265280.3
XP_005265337.1
28 - 29
4657
81
XM_005265282.3
XP_005265339.1
29 - 30
4799
81
XM_005265283.3
XP_005265340.1
28 - 29
4654
81
XM_011533900.2
XP_011532202.1
31 - 32
5006
81
XM_011533902.2
XP_011532204.1
30 - 31
4938
81
XM_011533903.2
XP_011532205.1
30 - 31
4943
81
XM_017006725.1
XP_016862214.1
30 - 31
4913
81
XM_017006726.1
XP_016862215.1
31 - 32
5023
81
XM_017006727.1
XP_016862216.1
31 - 32
4968
81
XM_017006728.1
XP_016862217.1
30 - 31
4964
81
XM_017006729.1
XP_016862218.1
30 - 31
5369
81
XM_017006730.1
XP_016862219.1
31 - 32
4959
81
XM_017006731.1
XP_016862220.1
30 - 31
4942
81
XM_017006732.1
XP_016862221.1
30 - 31
4935
81
XM_017006733.1
XP_016862222.1
30 - 31
5332
81
XM_017006734.1
XP_016862223.1
30 - 31
4905
81
XM_017006735.1
XP_016862224.1
29 - 30
5326
81
XM_017006736.1
XP_016862225.1
29 - 30
5324
81
XM_017006737.1
XP_016862226.1
29 - 30
5246
81
XM_017006738.1
XP_016862227.1
29 - 30
4867
81
XM_017006739.1
XP_016862228.1
29 - 30
5243
81
XM_017006740.1
XP_016862229.1
29 - 30
4784
81
XM_017006741.1
XP_016862230.1
29 - 30
4875
81
XM_017006742.1
XP_016862231.1
29 - 30
4898
81
XM_017006743.1
XP_016862232.1
30 - 31
5297
81
XM_017006744.1
XP_016862233.1
29 - 30
4864
81
XM_017006745.1
XP_016862234.1
29 - 30
5218
81
XM_017006746.1
XP_016862235.1
29 - 30
5215
81
XM_017006747.1
XP_016862236.1
29 - 30
5277
81
XM_017006748.1
XP_016862237.1
28 - 29
5173
81
XM_017006749.1
XP_016862238.1
28 - 29
4739
81
XM_017006750.1
XP_016862239.1
27 - 28
4627
81
XM_017006751.1
XP_016862240.1
29 - 30
4800
81
GenBank mRNA
AY281068.1
-
26 - 27
4455
81
Back To Top
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact
Tech Support
if you need more information.
Set Membership:
Amplicon length between 71 and 85
Homo sapiens
Probe spans exons
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
mitotic nuclear division
negative regulation of cell proliferation
covalent chromatin modification
DNA binding
chromatin binding
protein binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0