Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010832.hg.1 |
ENST00000368984.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010832.hg.1 |
ENST00000435263.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010832.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010832.hg.1 |
TC10000889.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000162712.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162712.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162712.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162712.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162712.hg.1 |
TC1000009152.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000368984.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
ENST00000420892.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009152.hg.1 |
NM_002775.4 |
Refseq |