Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009603.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009603.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009603.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009603.hg.1 |
TC10000802.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009604.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009604.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10009604.hg.1 |
TC10000802.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000160808.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160808.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160808.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160808.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160808.hg.1 |
TC1000008896.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008896.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160807.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160807.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160807.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160807.hg.1 |
ENST00000361804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160807.hg.1 |
ENST00000462899.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160807.hg.1 |
NM_005445.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160807.hg.1 |
TC1000008896.hg.1 |
Clariom D TC |