Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000460076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000460076.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023436.hg.1 |
TC01001516.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023436.hg.1 |
ENST00000239457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023436.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023436.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023437.hg.1 |
TC01001516.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023437.hg.1 |
ENST00000239457.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023437.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01023437.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178368.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178368.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178368.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178368.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178368.hg.1 |
TC0100010661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100178369.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178369.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178369.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100178369.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100178369.hg.1 |
TC0100010661.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000361951.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000460076.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000460076.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
ENST00000471476.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010661.hg.1 |
NM_018122.4 |
Refseq |