Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204466.1 | NP_001191395.1 | 18 - 19 | 2331 | 75 | |
NM_001204467.1 | NP_001191396.1 | 19 - 20 | 2559 | 75 | ||
NM_001204468.1 | NP_001191397.1 | 19 - 20 | 2593 | 75 | ||
NM_005676.4 | NP_005667.2 | 19 - 20 | 2562 | 75 | ||
NM_152856.2 | NP_690595.1 | 18 - 19 | 2328 | 75 | ||
XM_005272677.4 | XP_005272734.1 | 19 - 20 | 2612 | 75 | ||
XM_005272678.4 | XP_005272735.1 | 18 - 19 | 2146 | 75 | ||
XM_005272679.4 | XP_005272736.1 | 18 - 19 | 2143 | 75 | ||
XM_006724563.1 | XP_006724626.1 | 15 - 16 | 1779 | 75 | ||
XM_011543989.2 | XP_011542291.1 | 11 - 12 | 1503 | 75 | ||
XM_017029884.1 | XP_016885373.1 | 18 - 19 | 2482 | 75 | ||
XM_017029885.1 | XP_016885374.1 | 18 - 19 | 2254 | 75 | ||
GenBank mRNA | AK024839.1 | - | 2 - 3 | 75 | ||
AK025854.1 | - | 7 - 8 | 935 | 75 | ||
AK098115.1 | - | 1897 | 75 | |||
AK292758.1 | - | 18 - 19 | 2282 | 75 | ||
AL137421.1 | - | 20 - 21 | 2451 | 75 | ||
BC003089.1 | - | 18 - 19 | 2267 | 3536058 | 75 | |
BC004181.2 | - | 19 - 20 | 2515 | 2961094 | 75 | |
BC008733.2 | - | 19 - 20 | 2547 | 3163064 | 75 | |
BC024153.2 | - | 18 - 19 | 2177 | 3845258 | 75 | |
BX537969.1 | - | 19 - 20 | 2544 | 75 | ||
D50912.1 | - | 19 - 20 | 2410 | 75 | ||
KU178411.1 | - | 17 - 18 | 1935 | 75 | ||
KU178412.1 | - | 18 - 19 | 2169 | 75 | ||
U35373.1 | - | 5 - 6 | 546 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |