Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039245.hg.1 |
TC01002522.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039245.hg.1 |
ENST00000330210.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039245.hg.1 |
ENST00000373048.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039245.hg.1 |
ENST00000427468.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039245.hg.1 |
ENST00000446149.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039245.hg.1 |
ENST00000540011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01039245.hg.1 |
NM_001099439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
ENST00000373048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
ENST00000427468.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
ENST00000432874.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
ENST00000437645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
ENST00000446149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
ENST00000534097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
NM_001099439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
ENST00000373048.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
ENST00000427468.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
ENST00000432874.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
ENST00000437645.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
ENST00000446149.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
ENST00000534097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
NM_001099439.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100202635.hg.1 |
TC0100013752.hg.1 |
Clariom D TC |