Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000469889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000469889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000469889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000469889.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000496051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000496051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000496051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000496051.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000160.hg.1 |
ENST00000327451.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000160.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000160.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22000160.hg.1 |
TC22000024.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2200144564.hg.1 |
TC2200006520.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200144564.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144564.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144564.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144564.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144564.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144564.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144564.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144564.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200144564.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200144564.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000327451.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000399813.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000467228.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000469889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000469889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000469889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000469889.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000496051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000496051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000496051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000496051.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
ENST00000497401.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_001303484.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006520.hg.1 |
NM_031481.2 |
Refseq |