Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011787.hg.1 |
ENST00000215912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011787.hg.1 |
ENST00000402249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011787.hg.1 |
ENST00000416925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011787.hg.1 |
ENST00000441972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011787.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011787.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011787.hg.1 |
TC22000667.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011785.hg.1 |
ENST00000215912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011785.hg.1 |
ENST00000402249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011785.hg.1 |
ENST00000416925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011785.hg.1 |
ENST00000441972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011785.hg.1 |
ENST00000487265.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011785.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011785.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011785.hg.1 |
TC22000667.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011789.hg.1 |
ENST00000215912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011789.hg.1 |
ENST00000416925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011789.hg.1 |
ENST00000441972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011789.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011789.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011789.hg.1 |
TC22000667.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011784.hg.1 |
ENST00000215912.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011784.hg.1 |
ENST00000402249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011784.hg.1 |
ENST00000416925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011784.hg.1 |
ENST00000441972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011784.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011784.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22011784.hg.1 |
TC22000667.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160970.hg.1 |
TC2200008475.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160970.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160970.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160970.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160970.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160970.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160970.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160970.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160970.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160970.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160970.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160970.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160970.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160971.hg.1 |
TC2200008475.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160971.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160971.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160971.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160971.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160971.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160971.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160971.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160971.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160971.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160971.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160971.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160971.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160972.hg.1 |
TC2200008475.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160972.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160972.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160972.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160972.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160972.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160972.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160972.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160972.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160972.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160972.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160972.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160972.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160974.hg.1 |
TC2200008475.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160974.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160974.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160974.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160974.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160974.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160974.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160974.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160974.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000215912.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000402249.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000441972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000443175.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000480654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
ENST00000493034.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_001135911.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008475.hg.1 |
NM_052880.4 |
Refseq |