Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014682.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014682.hg.1 |
ENST00000558492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014682.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014682.hg.1 |
TC15001549.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014681.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014681.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014681.hg.1 |
TC15001549.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014687.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014687.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014687.hg.1 |
TC15001549.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014686.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014686.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15014686.hg.1 |
TC15001549.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500171909.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171909.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171909.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171909.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171909.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171909.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171909.hg.1 |
TC1500009736.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500171918.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171918.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171918.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171918.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171918.hg.1 |
TC1500009736.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500171910.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171910.hg.1 |
ENST00000558492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171910.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171910.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171910.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171910.hg.1 |
ENST00000558492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171910.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171910.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171910.hg.1 |
TC1500009736.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500171919.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171919.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171919.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171919.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171919.hg.1 |
TC1500009736.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500171911.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171911.hg.1 |
ENST00000558492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171911.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171911.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171911.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171911.hg.1 |
ENST00000558492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171911.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500171911.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500171911.hg.1 |
TC1500009736.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |