Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035258.1 | NP_001030335.1 | 1 - 2 | 52 | 135 | |
GenBank mRNA | BU532782.1 | - | 1 - 2 | 55 | 135 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |