Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000480529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000480529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000480529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000480529.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17002739.hg.1 |
ENST00000389022.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17002739.hg.1 |
ENST00000446264.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17002739.hg.1 |
ENST00000478373.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17002739.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17002739.hg.1 |
TC17000196.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000480529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000480529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000480529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000480529.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700007057.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
ENST00000389022.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
ENST00000470418.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
ENST00000478373.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
ENST00000483704.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
ENST00000582909.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
ENST00000616989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
NM_020201.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700149805.hg.1 |
TC1700007057.hg.1 |
Clariom D TC |