Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGOLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206841.1 | NP_001193770.1 | 3 - 4 | 114 | ||
GenBank mRNA | AF027515.1 | - | 3 - 4 | 1366 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |