Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001303512.1 | NP_001290441.1 | 2 - 3 | 577 | 62 | |
NM_001303515.1 | NP_001290444.1 | 2 - 3 | 412 | 62 | ||
GenBank mRNA | AK123822.1 | - | 2 - 3 | 412 | 62 | |
BC117277.1 | - | 2 - 3 | 369 | 40125828 | 62 | |
BC143545.1 | - | 2 - 3 | 368 | 9052056 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons |