Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377836.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377836.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377836.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377836.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377836.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000377837.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000471190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000471190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000487437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000487437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000487437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000487437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000487437.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033152.hg.1 |
TC01002150.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033152.hg.1 |
ENST00000377834.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01033152.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100193909.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100193909.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100193909.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100193909.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100193909.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100193909.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100193909.hg.1 |
TC0100012683.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377834.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377836.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377836.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377836.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377836.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377836.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000377837.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000471190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000471190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000487437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000487437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000487437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000487437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000487437.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
ENST00000489730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012683.hg.1 |
NM_019089.4 |
Refseq |