Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001830.hg.1 |
TC05000161.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001830.hg.1 |
ENST00000274254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001830.hg.1 |
ENST00000274255.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001830.hg.1 |
ENST00000308927.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001830.hg.1 |
ENST00000546211.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001830.hg.1 |
NM_001243120.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001830.hg.1 |
NM_005983.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05001830.hg.1 |
NM_032637.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000274254.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000274255.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000509692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000513151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000513263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000546211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000620197.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
NM_001243120.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
NM_005983.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
NM_032637.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000274254.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000274255.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000509692.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000513151.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000513263.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000546211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
ENST00000620197.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
NM_001243120.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
NM_005983.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
NM_032637.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148351.hg.1 |
TC0500007143.hg.1 |
Clariom D TC |