Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000427.hg.1 |
ENST00000354828.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000427.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000427.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000427.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21000427.hg.1 |
TC21000078.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000487266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000487266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000487266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000487266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000487266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
ENST00000487266.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000487266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000487266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000487266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000487266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000487266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
ENST00000487266.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2100006787.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100145798.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145798.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145798.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100145798.hg.1 |
NM_002040.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100145798.hg.1 |
TC2100006787.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100145798.hg.1 |
ENST00000354828.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145798.hg.1 |
ENST00000400075.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100145798.hg.1 |
NM_001197297.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100145798.hg.1 |
NM_002040.3 |
Refseq |