Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002430.hg.1 |
TC07000153.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002430.hg.1 |
ENST00000242152.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002430.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002430.hg.1 |
ENST00000407573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002430.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006941.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002427.hg.1 |
TC07000153.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002427.hg.1 |
ENST00000242152.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002427.hg.1 |
ENST00000407573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002427.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002431.hg.1 |
TC07000153.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002431.hg.1 |
ENST00000242152.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002431.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002431.hg.1 |
ENST00000407573.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002431.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700148063.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148063.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148063.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700148063.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148063.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148063.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700148063.hg.1 |
TC0700006941.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006941.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700148066.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148066.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148066.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148066.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700148066.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148066.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148066.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148066.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700148066.hg.1 |
TC0700006941.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700148067.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148067.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148067.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148067.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700148067.hg.1 |
ENST00000242152.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148067.hg.1 |
ENST00000405982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148067.hg.1 |
ENST00000407573.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700148067.hg.1 |
NM_000905.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700148067.hg.1 |
TC0700006941.hg.1 |
Clariom D TC |