Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018897.2 | NP_061720.2 | 27 - 28 | 4438 | 64 | |
XM_011511487.2 | XP_011509789.1 | 28 - 29 | 4814 | 64 | ||
XM_011511488.2 | XP_011509790.1 | 26 - 27 | 5488 | 64 | ||
XM_011511489.2 | XP_011509791.1 | 26 - 27 | 4400 | 64 | ||
XM_011511490.2 | XP_011509792.1 | 27 - 28 | 5608 | 64 | ||
XM_011511491.2 | XP_011509793.1 | 27 - 28 | 5610 | 64 | ||
XM_011511492.2 | XP_011509794.1 | 27 - 28 | 5610 | 64 | ||
XM_011511493.2 | XP_011509795.1 | 27 - 28 | 5610 | 64 | ||
XM_011511494.2 | XP_011509796.1 | 27 - 28 | 5609 | 64 | ||
XM_011511495.2 | XP_011509797.1 | 27 - 28 | 5609 | 64 | ||
XM_017004504.1 | XP_016859993.1 | 26 - 27 | 5334 | 64 | ||
GenBank mRNA | AB023161.2 | - | 27 - 28 | 4456 | 64 | |
AF327442.1 | - | 27 - 28 | 4433 | 64 | ||
Z83801.1 | - | 3 - 4 | 341 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |