Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021066.hg.1 |
TC12003279.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021066.hg.1 |
ENST00000317269.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021066.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021066.hg.1 |
ENST00000357606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021066.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021065.hg.1 |
TC12003279.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021065.hg.1 |
ENST00000317269.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021065.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021065.hg.1 |
ENST00000357606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021065.hg.1 |
ENST00000537962.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021065.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021069.hg.1 |
TC12003279.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021069.hg.1 |
ENST00000317269.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021069.hg.1 |
ENST00000357606.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021069.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203355.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203355.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203355.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203355.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203355.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203355.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203355.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203355.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203355.hg.1 |
TC1200012800.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200203356.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203356.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203356.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203356.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203356.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203356.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203356.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203356.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203356.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203356.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203356.hg.1 |
TC1200012800.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200203358.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203358.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203358.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203358.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203358.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203358.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203358.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203358.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203358.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203358.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203358.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203358.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203358.hg.1 |
TC1200012800.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200203362.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203362.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203362.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203362.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203362.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203362.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203362.hg.1 |
TC1200012800.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200012800.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203361.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203361.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203361.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203361.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203361.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203361.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203361.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203361.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203361.hg.1 |
TC1200012800.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200203357.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203357.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203357.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203357.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203357.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203357.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203357.hg.1 |
ENST00000317269.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203357.hg.1 |
ENST00000317287.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203357.hg.1 |
ENST00000357606.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203357.hg.1 |
ENST00000546957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203357.hg.1 |
ENST00000547445.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200203357.hg.1 |
NM_032364.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200203357.hg.1 |
TC1200012800.hg.1 |
Clariom D TC |