Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014835.3 | NP_055650.1 | 12 - 13 | 1452 | 57 | |
NM_144498.2 | NP_653081.1 | 12 - 13 | 1488 | 57 | ||
XM_017028165.1 | XP_016883654.1 | 13 - 14 | 1793 | 57 | ||
XM_017028167.1 | XP_016883656.1 | 13 - 14 | 1556 | 57 | ||
XM_017028170.1 | XP_016883659.1 | 12 - 13 | 1422 | 57 | ||
GenBank mRNA | AB018315.1 | - | 12 - 13 | 1358 | 57 | |
AF392447.1 | - | 12 - 13 | 1449 | 57 | ||
AK000230.1 | - | 12 - 13 | 1449 | 57 | ||
AK291851.1 | - | 12 - 13 | 1416 | 57 | ||
AK315576.1 | - | 12 - 13 | 1449 | 57 | ||
AY028168.1 | - | 12 - 13 | 1358 | 57 | ||
BC000296.2 | - | 12 - 13 | 1392 | 2819652 | 57 | |
BC004455.1 | - | 12 - 13 | 1392 | 2819652 | 57 | |
BC018812.2 | - | 12 - 13 | 1392 | 57 | ||
CR456722.1 | - | 11 - 12 | 1247 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |