Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290321.2 | NP_001277250.1 | 42 - 43 | 9179 | 110 | |
NM_001290322.2 | NP_001277251.1 | 41 - 42 | 10424 | 110 | ||
NM_005509.5 | NP_005500.4 | 41 - 42 | 9116 | 110 | ||
XM_005271909.4 | XP_005271966.1 | 44 - 45 | 8991 | 110 | ||
XM_005271910.4 | XP_005271967.1 | 42 - 43 | 9273 | 110 | ||
XM_005271912.2 | XP_005271969.1 | 44 - 45 | 8975 | 110 | ||
XM_011543212.2 | XP_011541514.1 | 43 - 44 | 9336 | 110 | ||
XM_011543213.2 | XP_011541515.1 | 44 - 45 | 8921 | 110 | ||
XM_011543214.2 | XP_011541516.1 | 40 - 41 | 8502 | 110 | ||
XM_011543215.2 | XP_011541517.1 | 39 - 40 | 8338 | 110 | ||
XM_017009141.1 | XP_016864630.1 | 43 - 44 | 8877 | 110 | ||
XM_017009142.1 | XP_016864631.1 | 42 - 43 | 8744 | 110 | ||
XM_017009143.1 | XP_016864632.1 | 45 - 46 | 9170 | 110 | ||
XM_017009144.1 | XP_016864633.1 | 43 - 44 | 8873 | 110 | ||
XM_017009145.1 | XP_016864634.1 | 43 - 44 | 8743 | 110 | ||
XM_017009146.1 | XP_016864635.1 | 43 - 44 | 10185 | 110 | ||
XM_017009148.1 | XP_016864637.1 | 42 - 43 | 9139 | 110 | ||
GenBank mRNA | AB593142.1 | - | 41 - 42 | 10427 | 110 | |
AK126726.1 | - | 3 - 4 | 1238 | 110 | ||
BC144537.1 | - | 42 - 43 | 8722 | 9053064 | 110 | |
BC150616.1 | - | 42 - 43 | 8722 | 9056986 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |