Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013894.hg.1 |
ENST00000335612.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013894.hg.1 |
ENST00000389720.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013894.hg.1 |
ENST00000389721.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013894.hg.1 |
ENST00000389722.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013894.hg.1 |
ENST00000389723.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013894.hg.1 |
ENST00000542895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013894.hg.1 |
ENST00000556626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013894.hg.1 |
NM_000347.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013894.hg.1 |
NM_001024858.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013894.hg.1 |
TC14001220.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
ENST00000389720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
ENST00000389721.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
ENST00000389722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
ENST00000553938.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
ENST00000556626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
NM_000347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
NM_001024858.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
ENST00000389720.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
ENST00000389721.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
ENST00000389722.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
ENST00000553938.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
ENST00000556626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
NM_000347.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
NM_001024858.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400169569.hg.1 |
TC1400009417.hg.1 |
Clariom D TC |