Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011542984.2 | XP_011541286.1 | 1 | 226 | 65 | |
XM_017018305.1 | XP_016873794.1 | 1 | 226 | 65 | ||
XM_017018306.1 | XP_016873795.1 | 1 | 226 | 65 | ||
XM_017018308.1 | XP_016873797.1 | 1 | 226 | 65 | ||
XM_017018311.1 | XP_016873800.1 | 1 | 226 | 65 | ||
XM_017018312.1 | XP_016873801.1 | 1 | 226 | 65 | ||
GenBank mRNA | AF161533.1 | - | 1 | 658 | 65 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mastermind like transcriptional coactivator 2 | MAML2 |
MAM-3, MAM2, MAM3, MLL-MAML2 |
-45.5kb | Sense | Chr. 11 - 95976593 - 96343245 on NCBI Build GRCh38 |
Downstream | JRK-like | JRKL | HHMJG | 560.0 | Anti-sense | Chr. 11 - 96389994 - 96393563 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |