Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000559079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000559079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000559079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000559079.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006307.hg.1 |
ENST00000388942.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006307.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15006307.hg.1 |
TC15000706.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1500156735.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156735.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156735.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156735.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156735.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156735.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156735.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156735.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156735.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500156735.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500156735.hg.1 |
TC1500007914.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000388942.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000484722.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000558249.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000559079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000559079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000559079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000559079.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
ENST00000561302.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007914.hg.1 |
NM_152335.2 |
Refseq |