Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000460442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000460442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000460442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000460442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000460442.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000497422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000497422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000497422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
ENST00000497422.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300202272.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202272.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202272.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202272.hg.1 |
ENST00000460442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202272.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202272.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300202272.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202272.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202272.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202272.hg.1 |
ENST00000460442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202272.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300202272.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300202272.hg.1 |
TC0300013696.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000296326.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000397544.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000438232.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000460442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000460442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000460442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000460442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000460442.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000465519.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000488508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000497422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000497422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000497422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
ENST00000497422.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300013696.hg.1 |
NM_001039617.1 |
Refseq |