Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000600762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000600762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000600762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000600762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000602234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000602234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000602234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000602234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
ENST00000602234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017816.hg.1 |
TC19002686.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017816.hg.1 |
ENST00000269881.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19017816.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000600762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000600762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000600762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000600762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000602234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000602234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000602234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000602234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
ENST00000602234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011899.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204945.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204945.hg.1 |
ENST00000600762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204945.hg.1 |
ENST00000602234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204945.hg.1 |
NM_145046.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900204945.hg.1 |
TC1900011899.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900204945.hg.1 |
ENST00000269881.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204945.hg.1 |
ENST00000600762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204945.hg.1 |
ENST00000602234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900204945.hg.1 |
NM_145046.4 |
Refseq |