Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000389266.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000444666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000444666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000444666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000444666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000444666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000444666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000444666.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000454308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000454308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000454308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000454308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000454308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000454308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000454308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000454308.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000465748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000465748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000465748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000465748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000465748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000465748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000470392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000470392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000470392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000470392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000478124.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000484093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000484093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000484093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000484093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000484093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000485784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000485784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000485784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000485784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000485784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000485784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000496643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000496643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000496643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000496643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000496643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000496643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000627489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000627489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
ENST00000627489.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700007098.hg.1 |
NM_002047.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700011186.hg.1 |
ENST00000570895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700011186.hg.1 |
ENST00000570895.1 |
Ensembl |