Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022034.hg.1 |
TC05001614.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022034.hg.1 |
ENST00000311106.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022034.hg.1 |
ENST00000508626.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022034.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022034.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000509190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000509190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000509190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000509190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000509190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500179799.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500179799.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500179799.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500179799.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500179799.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500179799.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500179799.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500179799.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500179799.hg.1 |
TC0500011513.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000311106.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000508626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000509190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000509190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000509190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000509190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000509190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
ENST00000513085.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_000439.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011513.hg.1 |
NM_001177875.1 |
Refseq |