Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042993.hg.1 |
TC02002673.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042993.hg.1 |
ENST00000286190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042993.hg.1 |
ENST00000392257.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042993.hg.1 |
ENST00000405148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042993.hg.1 |
ENST00000439709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042993.hg.1 |
ENST00000448967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042993.hg.1 |
NM_001127391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042993.hg.1 |
NM_139163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042994.hg.1 |
TC02002673.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042994.hg.1 |
ENST00000286190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042994.hg.1 |
ENST00000392257.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042994.hg.1 |
ENST00000405148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042994.hg.1 |
ENST00000439709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042994.hg.1 |
ENST00000448967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042994.hg.1 |
NM_001127391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042994.hg.1 |
NM_139163.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042991.hg.1 |
TC02002673.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042991.hg.1 |
ENST00000286190.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042991.hg.1 |
ENST00000392257.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042991.hg.1 |
ENST00000405148.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042991.hg.1 |
ENST00000439709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042991.hg.1 |
ENST00000448967.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042991.hg.1 |
NM_001127391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02042991.hg.1 |
NM_139163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
ENST00000392257.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
ENST00000405148.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
ENST00000439709.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
ENST00000494171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
NM_001127391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
NM_001289993.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
NM_139163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
NR_110620.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
ENST00000392257.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
ENST00000405148.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
ENST00000439709.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
ENST00000494171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
NM_001127391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
NM_001289993.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
NM_139163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
NR_110620.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213160.hg.1 |
TC0200015424.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
ENST00000392257.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
ENST00000405148.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
ENST00000439709.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
NM_001127391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
NM_001289993.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
NM_139163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
NR_110620.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
ENST00000392257.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
ENST00000405148.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
ENST00000439709.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
NM_001127391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
NM_001289993.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
NM_139163.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
NR_110620.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200213161.hg.1 |
TC0200015424.hg.1 |
Clariom D TC |