Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007528.hg.1 |
TC03000343.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007528.hg.1 |
ENST00000233025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007528.hg.1 |
ENST00000423431.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03007528.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000448693.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000448693.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000448693.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000448693.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000448693.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000448693.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000448693.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000448693.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000448693.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000448693.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300007495.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
ENST00000448693.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
ENST00000233025.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
ENST00000423431.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
ENST00000448693.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
ENST00000474945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
ENST00000602728.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
ENST00000619898.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
NM_014041.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300154135.hg.1 |
TC0300007495.hg.1 |
Clariom D TC |