Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000555333.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006300.hg.1 |
ENST00000238561.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006300.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14006300.hg.1 |
TC14000494.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000341211.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555217.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000555333.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000556560.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_001142545.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400007769.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155231.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155231.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155231.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155231.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155231.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155231.hg.1 |
ENST00000238561.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155231.hg.1 |
ENST00000393639.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155231.hg.1 |
ENST00000556048.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155231.hg.1 |
ENST00000557501.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400155231.hg.1 |
NM_020421.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400155231.hg.1 |
TC1400007769.hg.1 |
Clariom D TC |