Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013351.hg.1 |
ENST00000339762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013351.hg.1 |
ENST00000554729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013351.hg.1 |
ENST00000556826.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013351.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14013351.hg.1 |
TC14001172.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
ENST00000557430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
ENST00000556306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
ENST00000556306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400182449.hg.1 |
TC1400010752.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000339762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000554729.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000556826.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557254.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
ENST00000557430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001206920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010752.hg.1 |
NM_001306087.1 |
Refseq |