Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs01027251_m1
See other DTNB GE Assays ›
Gene Symbol:
DTNB
Gene Name:
dystrobrevin beta
Gene Aliases:
-
Chromosome Location:
Chr.2: 25377220 - 25673647 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs01027251_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: DTNB
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001320934.1
NP_001307863.1
14 - 15
1725
65
NM_001320936.1
NP_001307865.1
15 - 16
1815
65
NM_021907.4
NP_068707.1
15 - 16
1815
65
NM_033147.3
NP_149159.2
15 - 16
1815
65
NM_033148.3
NP_149160.1
14 - 15
1725
65
NM_183360.2
NP_899204.1
15 - 16
1815
65
XM_011532671.1
XP_011530973.1
15 - 16
1750
65
XM_011532672.2
XP_011530974.1
15 - 16
1750
65
XM_011532674.1
XP_011530976.1
15 - 16
1750
65
XM_011532676.1
XP_011530978.1
14 - 15
1660
65
XM_011532677.2
XP_011530979.1
14 - 15
2206
65
XM_011532680.2
XP_011530982.1
14 - 15
1671
65
XM_011532681.2
XP_011530983.1
14 - 15
1646
65
XM_017003532.1
XP_016859021.1
14 - 15
1660
65
XM_017003533.1
XP_016859022.1
14 - 15
1642
65
XM_017003537.1
XP_016859026.1
11 - 12
2103
65
XM_017003539.1
XP_016859028.1
13 - 14
1517
65
XM_017003540.1
XP_016859029.1
13 - 14
1556
65
XM_017003541.1
XP_016859030.1
13 - 14
1540
65
XM_017003542.1
XP_016859031.1
13 - 14
1512
65
XM_017003543.1
XP_016859032.1
12 - 13
1406
65
XM_017003544.1
XP_016859033.1
13 - 14
1494
65
XM_017003545.1
XP_016859034.1
12 - 13
1388
65
GenBank mRNA
AF022728.1
-
15 - 16
1713
65
AF070567.1
-
9 - 10
957
65
AK094494.1
-
14 - 15
1533
65
BC049366.1
-
15 - 16
1806
4817552
65
Y12712.1
-
14 - 15
1645
65
Y15722.1
-
14 - 15
1552
65
Back To Top
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact
Tech Support
if you need more information.
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene DTNB
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0