Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059471.hg.1 |
TC01003901.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059471.hg.1 |
ENST00000272117.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059471.hg.1 |
ENST00000429204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01059471.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000366784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000366784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000366784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000366784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000366784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000366784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000366784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000366784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000366784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000366784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017534.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232708.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232708.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232708.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232708.hg.1 |
ENST00000272117.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232708.hg.1 |
ENST00000429204.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100232708.hg.1 |
NM_002221.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100232708.hg.1 |
TC0100017534.hg.1 |
Clariom D TC |