Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899564 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048551 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899559 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899571 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899527 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899560 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899597 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899568 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048464 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899540 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899599 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048577 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048557 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048528 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899534 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048472 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048590 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899537 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048597 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15538399 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899555 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048603 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048574 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899557 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048555 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899549 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899535 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899566 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15538435 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899600 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899531 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899563 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048568 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899593 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899583 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048599 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048547 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048571 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899577 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048591 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048560 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899581 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899576 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P15538449 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899550 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899570 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P02899525 |
- |
- |
Agilent Human Whole Genome ChIP Tiling Array |
A_17_P23048655 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
11258 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
172203 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 4x180K |
159873 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 8x60K |
18341 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
925156 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
769312 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
425328 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
802130 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
511038 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
707318 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
543278 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
266660 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
234848 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
907372 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
628965 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
151110 |
- |
- |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M |
326220 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
391394 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
163380 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
9293 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
311379 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
54353 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
375572 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
419089 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
227293 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
158891 |
AK311193.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
158891 |
BC114549.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
158891 |
NM_000829.3 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
158891 |
NM_001077243.2 |
refseq_mrna_human_NM |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
158891 |
U16129.1 |
genbank_mrna_human |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
327426 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
89660 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
353386 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
37970 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
18410 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
351576 |
- |
- |
Agilent SurePrint G3 Human CNV Microarray 2x400K |
409689 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3294821 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P306307 |
- |
- |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k |
GT_44k_23_P306307 |
- |
- |
Agilent Whole Human Genome Oligo Microarray 4x44K |
29279 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P103173 |
- |
- |
Agilent Human Genome CGH Microarray 44B |
A_14_P200706 |
- |
- |
Agilent Human Genome CGH Microarray 44A |
A_14_P103173 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
54840 |
- |
- |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray |
29135 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_18_P14582504 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P00910532 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P00910523 |
- |
- |
Agilent Human Genome CGH + SNP Microarray |
A_16_P31224758 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_23_P306307 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K v2 |
A_33_P3294821 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_23_P306307 |
- |
- |
SurePrint G3 Human GE Microarray G4851A |
A_33_P3294821 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15538484 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899594 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23048655 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899597 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899596 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899591 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899598 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23048653 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899599 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23048669 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899600 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899593 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899601 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899600 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899599 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23048669 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899591 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899597 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899598 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899594 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899596 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899601 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23048655 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P23048653 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P15538484 |
- |
- |
Agilent SurePrint G3 Human Promoter Microarray 1x1M |
A_17_P02899593 |
- |
- |
Whole Human Genome Oligo Microarray G4112A |
A_23_P306307 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P103173 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_16_P00910520 |
- |
- |
Agilent Human Genome CGH Microarray 105A (G4412A) |
A_14_P200706 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02899597 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02899592 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02899595 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02899596 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02899591 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02899600 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02899599 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02899594 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02899590 |
- |
- |
Agilent Human Promoter ChIP-on-Chip A) |
A_17_P02899598 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15538461 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15538464 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15538477 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15538469 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15538476 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15538125 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P15538475 |
- |
- |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) |
A_17_P02899594 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P103173 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_14_P200706 |
- |
- |
Agilent Human Genome 244K CGH Microarray (Alpha Test) |
A_16_P00910520 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P200706 |
- |
- |
Agilent Human Genome CGH Microarray 4x44K |
A_14_P103173 |
- |
- |
Agilent Whole Human Genome Microarray 4x44K G4112F |
A_23_P306307 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
150850 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
43082 |
- |
- |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 |
68302 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
86593 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
29059 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
59848 |
- |
- |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 |
27170 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P14585212 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P14582504 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P14582543 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P00910523 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P36623480 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P31224758 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P16573294 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_16_P31224703 |
- |
- |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) |
A_18_P14579577 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899587 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899572 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899555 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899569 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899567 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899535 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15538464 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899588 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899545 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15538461 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899583 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899565 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899547 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899574 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899526 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899553 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899562 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899594 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899591 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899566 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899598 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899530 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899560 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899527 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15538477 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P15538359 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899600 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899551 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899576 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899540 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899541 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899549 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899559 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899537 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899556 |
- |
- |
Agilent Human Whole Genome ChIP-on-Chip |
A_17_P02899542 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P16573279 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P16573309 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_18_P14582543 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P00910523 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_16_P16573293 |
- |
- |
Agilent SurePrint G3 Human CGH Microarray 2x400K |
A_18_P14582504 |
- |
- |