Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000504941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000504941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000504941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000504941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000504941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000505499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000505499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000509503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000509503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000509503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000509503.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000510359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000510359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000510359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000510359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000510359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022451.hg.1 |
TC05001657.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022451.hg.1 |
ENST00000333274.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022451.hg.1 |
ENST00000510359.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05022451.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180578.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180578.hg.1 |
ENST00000510359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180578.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180578.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180578.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180578.hg.1 |
ENST00000510359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180578.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500180578.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500180578.hg.1 |
TC0500011648.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000333274.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000504941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000504941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000504941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000504941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000504941.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000505499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000505499.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000509503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000509503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000509503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000509503.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000510359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000510359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000510359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000510359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000510359.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
ENST00000611503.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011648.hg.1 |
NM_001962.2 |
Refseq |