Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014213.hg.1 |
TC16002064.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014213.hg.1 |
ENST00000333496.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014213.hg.1 |
ENST00000354630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014213.hg.1 |
ENST00000355477.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014213.hg.1 |
ENST00000357076.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014213.hg.1 |
ENST00000357806.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014213.hg.1 |
ENST00000357857.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014213.hg.1 |
ENST00000359984.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014213.hg.1 |
ENST00000360019.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014213.hg.1 |
ENST00000395653.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014213.hg.1 |
ENST00000535392.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014213.hg.1 |
NM_000086.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16014213.hg.1 |
NM_001042432.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000333496.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000355477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000357806.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000357857.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000359984.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000360019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000395653.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000561689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000563874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000564574.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000565047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000565140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000565316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000565354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000565778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000566057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000566083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000566824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000567495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000567963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000569030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000569430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000623866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000628023.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000631023.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
NM_000086.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
NM_001042432.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
NM_001286104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
NM_001286105.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
NM_001286109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
NM_001286110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000333496.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000355477.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000357806.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000357857.13 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000359984.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000360019.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000395653.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000561689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000563874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000564574.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000565047.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000565140.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000565316.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000565354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000565778.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000566057.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000566083.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000566824.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000567495.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000567963.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568076.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568224.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568422.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568452.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568472.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568497.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000568558.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000569030.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000569430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000623866.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000628023.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
ENST00000631023.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
NM_000086.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
NM_001042432.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
NM_001286104.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
NM_001286105.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
NM_001286109.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
NM_001286110.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600175859.hg.1 |
TC1600009861.hg.1 |
Clariom D TC |