Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000622957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000622957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
ENST00000622957.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024960.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024960.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11024960.hg.1 |
TC11002081.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100189668.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189668.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189668.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100189668.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100189668.hg.1 |
TC1100011611.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000298198.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000622957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000622957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
ENST00000622957.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100011611.hg.1 |
NM_173582.4 |
Refseq |