Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000419693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000419693.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000480807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000480807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000480807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000480807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000480807.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000494722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000494722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000494722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000494722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
ENST00000617724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013502.hg.1 |
NR_106895.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07016889.hg.1 |
TC07001096.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07016889.hg.1 |
ENST00000222990.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07016889.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700199736.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700199736.hg.1 |
ENST00000480807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700199736.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700199736.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700199736.hg.1 |
ENST00000480807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700199736.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700199736.hg.1 |
TC0700013502.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000222990.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000419693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000419693.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000435060.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000457286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000479689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000480807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000480807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000480807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000480807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000480807.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000494722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000494722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000494722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000494722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
ENST00000617724.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NM_013321.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013502.hg.1 |
NR_106895.1 |
Refseq |