Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036068.hg.1 |
TC02002220.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036068.hg.1 |
ENST00000259213.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02036068.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202116.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202116.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202116.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202116.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202116.hg.1 |
TC0200013922.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
ENST00000327407.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013922.hg.1 |
NM_173178.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202117.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202117.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202117.hg.1 |
ENST00000259213.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200202117.hg.1 |
NM_014438.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200202117.hg.1 |
TC0200013922.hg.1 |
Clariom D TC |