Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000451518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000451518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000451518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000451518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000451518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000483555.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000483555.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000483555.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000483555.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000483555.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000483555.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000495597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000495597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000495597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000495597.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000498552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
ENST00000498552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000393951.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000404568.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000418139.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000451518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000451518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000451518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000451518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000451518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000473508.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000483555.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000483555.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000483555.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000483555.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000483555.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000483555.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000495597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000495597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000495597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000495597.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000498552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
ENST00000498552.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013111.hg.1 |
NM_133637.2 |
Refseq |