Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005270028.3 | XP_005270085.1 | 1 - 2 | 175 | 88 | |
XM_011540060.2 | XP_011538362.1 | 1 - 2 | 175 | 88 | ||
XM_017016514.1 | XP_016872003.1 | 1 - 2 | 175 | 88 | ||
GenBank mRNA | AK027673.1 | - | 1 - 2 | 109 | 88 | |
BC009738.1 | - | 1 - 2 | 123 | 3879159 | 88 | |
GQ472233.1 | - | 1 - 2 | 123 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens 1700 Probe spans exons |