Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199282.2 | NP_001186211.2 | 35 - 36 | 5934 | 104 | |
NM_006726.4 | NP_006717.2 | 35 - 36 | 5893 | 104 | ||
XM_005263372.3 | XP_005263429.1 | 35 - 36 | 6079 | 104 | ||
XM_005263373.2 | XP_005263430.1 | 35 - 36 | 6121 | 104 | ||
XM_005263374.3 | XP_005263431.1 | 35 - 36 | 6078 | 104 | ||
XM_005263375.3 | XP_005263432.1 | 35 - 36 | 6078 | 104 | ||
XM_011532434.2 | XP_011530736.1 | 35 - 36 | 6078 | 104 | ||
XM_017008872.1 | XP_016864361.1 | 35 - 36 | 6121 | 104 | ||
GenBank mRNA | AB208838.1 | - | 11 - 12 | 1607 | 104 | |
AF216648.2 | - | 35 - 36 | 5893 | 104 | ||
AF467287.1 | - | 35 - 36 | 5934 | 104 | ||
M83822.1 | - | 17 - 18 | 3329 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |