Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321312.1 | NP_001308241.1 | 5 - 6 | 1406 | 74 | |
NM_001321313.1 | NP_001308242.1 | 5 - 6 | 1192 | 74 | ||
NM_005966.3 | NP_005957.2 | 7 - 8 | 1668 | 74 | ||
XM_005246582.1 | XP_005246639.1 | 6 - 7 | 1341 | 74 | ||
XM_005246583.1 | XP_005246640.1 | 6 - 7 | 1455 | 74 | ||
XM_011511219.2 | XP_011509521.1 | 6 - 7 | 1310 | 74 | ||
XM_017004170.1 | XP_016859659.1 | 6 - 7 | 1351 | 74 | ||
XM_017004176.1 | XP_016859665.1 | 5 - 6 | 1260 | 74 | ||
GenBank mRNA | AF045451.1 | - | 4 - 5 | 1115 | 74 | |
AK292446.1 | - | 5 - 6 | 1206 | 74 | ||
BC035724.1 | - | 7 - 8 | 1557 | 5721972 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |