Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
ENST00000401179.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900008827.hg.1 |
NR_030632.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900168729.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900168729.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900168729.hg.1 |
TC1900008827.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900168729.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900168729.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
ENST00000270458.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
ENST00000401179.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
NM_031895.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900008827.hg.1 |
NR_030632.1 |
Refseq |