Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008817.hg.1 |
ENST00000300086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008817.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008817.hg.1 |
TC16000621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008816.hg.1 |
ENST00000300086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008816.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008816.hg.1 |
TC16000621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008813.hg.1 |
ENST00000300086.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008813.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16008813.hg.1 |
TC16000621.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
ENST00000569234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
ENST00000569234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
ENST00000569234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
ENST00000569234.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006518.hg.1 |
ENST00000455617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191459.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191459.hg.1 |
ENST00000569234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191459.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191459.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191459.hg.1 |
ENST00000569234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191459.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191459.hg.1 |
TC1600011428.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600191454.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191454.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191454.hg.1 |
TC1600011428.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600191461.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191461.hg.1 |
ENST00000569234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191461.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191461.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191461.hg.1 |
ENST00000569234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191461.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191461.hg.1 |
TC1600011428.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
ENST00000569234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
ENST00000569234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
ENST00000569234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
ENST00000569234.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011428.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006518.hg.1 |
ENST00000455617.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191457.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191457.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191457.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191457.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191457.hg.1 |
TC1600011428.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600191458.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191458.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191458.hg.1 |
ENST00000300086.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191458.hg.1 |
NM_018975.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191458.hg.1 |
TC1600011428.hg.1 |
Clariom D TC |